Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RB1
Entrez Gene ID
5925
Gene Name
RB transcriptional corepressor 1
Gene Aliases
OSRC, PPP1R130, RB, p105-Rb, pRb, pp110
Chromosome Location
Chr.13: 48303747 - 48481890 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000321.2 NP_000312.2 26 - 27 2877 68
XM_011535171.2 XP_011533473.1 25 - 26 2466 68
GenBank mRNA AB208788.1 - 26 - 27 2808 68
AK291258.1 - 26 - 27 2898 68
AK299179.1 - 27 - 28 2788 68
AK300284.1 - 27 - 28 2814 68
AK307125.1 - 2 - 3 432 68
BC039060.1 - 26 - 27 2863 68
BC040540.1 - 26 - 27 2873 68
L41870.1 - 26 - 27 2848 68
M15400.1 - 26 - 27 2849 68
M28419.1 - 26 - 27 2849 68
M33647.1 - 26 - 27 2714 68

View Full Product Details