Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RB1
Entrez Gene ID
5925
Gene Name
RB transcriptional corepressor 1
Gene Aliases
OSRC, PPP1R130, RB, p105-Rb, pRb, pp110
Chromosome Location
Chr.13: 48303747 - 48481890 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000321.2 NP_000312.2 6 - 7 766 142
XM_011535171.2 XP_011533473.1 5 - 6 355 142
GenBank mRNA AB208788.1 - 6 - 7 697 142
AK291258.1 - 6 - 7 787 142
AK299179.1 - 7 - 8 677 142
AK300284.1 - 7 - 8 703 142
AK307730.1 - 6 - 7 142
BC039060.1 - 6 - 7 752 142
BC040540.1 - 6 - 7 762 142
L41870.1 - 6 - 7 737 142
M15400.1 - 6 - 7 738 142
M28419.1 - 6 - 7 738 142
M33647.1 - 6 - 7 603 142

View Full Product Details