The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022066.3 | NP_071349.3 | 7 - 8 | 1068 | 72 |
XM_005257567.1 | XP_005257624.1 | 7 - 8 | 1052 | 72 | |
XM_017024934.1 | XP_016880423.1 | 7 - 8 | 1052 | 72 | |
XM_017024935.1 | XP_016880424.1 | 7 - 8 | 1052 | 72 | |
GenBank mRNA | AB051521.1 | - | 7 - 8 | 1068 | 72 |
BC036820.1 | - | 2 - 3 | 204 | 72 |