The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005498.3 | NP_001005498.2 | 8 - 9 | 1331 | 85 |
NM_024599.5 | NP_078875.4 | 9 - 10 | 1493 | 85 | |
XM_005257669.3 | XP_005257726.1 | 9 - 10 | 1854 | 85 | |
XM_005257670.1 | XP_005257727.1 | 9 - 10 | 1363 | 85 | |
XM_011525249.2 | XP_011523551.1 | 9 - 10 | 2810 | 85 | |
XM_011525250.2 | XP_011523552.1 | 10 - 11 | 1520 | 85 | |
XM_011525251.2 | XP_011523553.1 | 9 - 10 | 1337 | 85 | |
XM_017025079.1 | XP_016880568.1 | 9 - 10 | 1767 | 85 | |
GenBank mRNA | AK025994.1 | - | 4 - 5 | 517 | 85 |
AK074034.1 | - | 7 - 8 | 1798 | 85 | |
AK225684.1 | - | 4 - 5 | 470 | 85 | |
AK292135.1 | - | 9 - 10 | 1486 | 85 | |
AK292166.1 | - | 8 - 9 | 1286 | 85 | |
AK303434.1 | - | 7 - 8 | 1147 | 85 | |
AL833680.1 | - | 3 - 4 | 386 | 85 | |
AY500247.1 | - | 8 - 9 | 1182 | 85 | |
BC016034.1 | - | 9 - 10 | 1473 | 85 | |
BC035829.1 | - | 4 - 5 | 579 | 85 |