The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005498.3 | NP_001005498.2 | 12 - 13 | 1792 | 99 |
NM_024599.5 | NP_078875.4 | 13 - 14 | 1954 | 99 | |
XM_005257669.3 | XP_005257726.1 | 13 - 14 | 2315 | 99 | |
XM_005257670.1 | XP_005257727.1 | 13 - 14 | 1824 | 99 | |
XM_011525249.2 | XP_011523551.1 | 13 - 14 | 3271 | 99 | |
XM_011525250.2 | XP_011523552.1 | 14 - 15 | 1981 | 99 | |
XM_011525251.2 | XP_011523553.1 | 13 - 14 | 1798 | 99 | |
XM_017025079.1 | XP_016880568.1 | 13 - 14 | 2228 | 99 | |
GenBank mRNA | AK025994.1 | - | 8 - 9 | 978 | 99 |
AK074034.1 | - | 11 - 12 | 2259 | 99 | |
AK225684.1 | - | 8 - 9 | 931 | 99 | |
AK292166.1 | - | 12 - 13 | 1747 | 99 | |
AL833680.1 | - | 7 - 8 | 847 | 99 | |
AY500247.1 | - | 12 - 13 | 1643 | 99 | |
BC016034.1 | - | 13 - 14 | 1934 | 99 | |
BC035829.1 | - | 8 - 9 | 1040 | 99 | |
BE076010.1 | - | 2 - 3 | 99 |