The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001173516.1 | NP_001166987.1 | 9 - 10 | 1655 | 58 |
NM_001173517.1 | NP_001166988.1 | 8 - 9 | 1662 | 58 | |
NM_024597.3 | NP_078873.2 | 9 - 10 | 1767 | 58 | |
XM_017029843.1 | XP_016885332.1 | 9 - 10 | 1619 | 58 | |
GenBank mRNA | AK301478.1 | - | 9 - 10 | 1656 | 58 |
AL832120.1 | - | 8 - 9 | 1454 | 58 | |
BC040518.1 | - | 9 - 10 | 1631 | 58 | |
BC064350.1 | - | 9 - 10 | 1447 | 58 |