The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018303.5 | NP_060773.3 | 1 - 2 | 121 | 80 |
XM_017011022.1 | XP_016866511.1 | 1 - 2 | 121 | 80 | |
XM_017011023.1 | XP_016866512.1 | 1 - 2 | 121 | 80 | |
XM_017011025.1 | XP_016866514.1 | 1 - 2 | 121 | 80 | |
XM_017011026.1 | XP_016866515.1 | 1 - 2 | 121 | 80 | |
GenBank mRNA | AJ414403.1 | - | 1 - 2 | 45 | 80 |
AK056564.1 | - | 1 - 2 | 89 | 80 | |
AK314628.1 | - | 1 - 2 | 121 | 80 | |
AL833213.1 | - | 1 - 2 | 67 | 80 | |
DA228027.1 | - | 1 - 2 | 121 | 80 |