The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003472.3 | NP_003463.1 | 3 - 4 | 436 | 152 |
XM_017011273.1 | XP_016866762.1 | 3 - 4 | 408 | 152 | |
GenBank mRNA | AK298089.1 | - | 3 - 4 | 348 | 152 |
AK312616.1 | - | 3 - 4 | 365 | 152 | |
AU131262.1 | - | 3 - 4 | 436 | 152 | |
BC035259.1 | - | 3 - 4 | 408 | 152 | |
X64229.1 | - | 3 - 4 | 276 | 152 |