The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006051.3 | NP_006042.3 | 3 - 4 | 647 | 139 |
NM_133172.2 | NP_573418.2 | 3 - 4 | 647 | 139 | |
NM_133173.2 | NP_573419.2 | 3 - 4 | 647 | 139 | |
NM_133174.2 | NP_573420.2 | 3 - 4 | 647 | 139 | |
GenBank mRNA | AB018247.1 | - | 3 - 4 | 339 | 139 |
AB049618.1 | - | 3 - 4 | 375 | 139 | |
AB061224.1 | - | 3 - 4 | 288 | 139 | |
AB079687.1 | - | 3 - 4 | 288 | 139 | |
AB094990.1 | - | 3 - 4 | 288 | 139 | |
AB209839.1 | - | 3 - 4 | 375 | 139 | |
AF224708.1 | - | 3 - 4 | 308 | 139 | |
AF224709.1 | - | 3 - 4 | 308 | 139 | |
AF224710.1 | - | 3 - 4 | 308 | 139 | |
AF224711.1 | - | 3 - 4 | 308 | 139 | |
AK075280.1 | - | 3 - 4 | 610 | 139 | |
AK125244.1 | - | 2 - 3 | 908 | 139 | |
BC013158.1 | - | 3 - 4 | 647 | 139 | |
BC125187.1 | - | 3 - 4 | 639 | 139 |