The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006051.3 | NP_006042.3 | 9 - 10 | 1210 | 71 |
NM_133172.2 | NP_573418.2 | 8 - 9 | 1204 | 71 | |
NM_133173.2 | NP_573419.2 | 9 - 10 | 1189 | 71 | |
NM_133174.2 | NP_573420.2 | 8 - 9 | 1183 | 71 | |
GenBank mRNA | AB018247.1 | - | 9 - 10 | 881 | 71 |
AB049618.1 | - | 9 - 10 | 1073 | 71 | |
AB061224.1 | - | 9 - 10 | 976 | 71 | |
AB079687.1 | - | 9 - 10 | 830 | 71 | |
AB094990.1 | - | 8 - 9 | 824 | 71 | |
AB209839.1 | - | 9 - 10 | 917 | 71 | |
AF224708.1 | - | 8 - 9 | 865 | 71 | |
AF224709.1 | - | 9 - 10 | 850 | 71 | |
AF224710.1 | - | 8 - 9 | 844 | 71 | |
AF224711.1 | - | 9 - 10 | 871 | 71 | |
AK074885.1 | - | 4 - 5 | 1116 | 71 | |
AK075280.1 | - | 9 - 10 | 1152 | 71 | |
AK125244.1 | - | 6 - 7 | 1842 | 71 | |
AK304824.1 | - | 5 - 6 | 1210 | 71 | |
BC013158.1 | - | 9 - 10 | 1189 | 71 | |
BC125187.1 | - | 9 - 10 | 1181 | 71 |