Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAAF1
Entrez Gene ID
123872
Gene Name
dynein axonemal assembly factor 1
Gene Aliases
CILD13, LRRC50, ODA7
Chromosome Location
Chr.16: 84145260 - 84178761 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318756.1 NP_001305685.1 2 - 3 297 93
NM_178452.5 NP_848547.4 6 - 7 1047 93
XM_006721129.2 XP_006721192.1 6 - 7 1047 93
XM_011522853.2 XP_011521155.1 6 - 7 1047 93
XM_011522854.2 XP_011521156.1 6 - 7 1047 93
XM_011522855.2 XP_011521157.1 6 - 7 1047 93
XM_011522857.2 XP_011521159.1 6 - 7 1047 93
XM_011522858.2 XP_011521160.1 6 - 7 1047 93
XM_017022918.1 XP_016878407.1 6 - 7 1047 93
XM_017022919.1 XP_016878408.1 6 - 7 1355 93
XM_017022920.1 XP_016878409.1 2 - 3 495 93
XM_017022921.1 XP_016878410.1 2 - 3 495 93
XM_017022922.1 XP_016878411.1 2 - 3 495 93
GenBank mRNA AK057238.1 - 6 - 7 999 93
AK058059.1 - 2 - 3 304 93
AK295990.1 - 2 - 3 297 93
AL833327.1 - 6 - 7 1018 93
AL833336.1 - 6 - 7 1018 93
BC024009.2 - 6 - 7 990 93
BX648657.1 - 6 - 7 1020 93

View Full Product Details