The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291860.1 | NP_001278789.1 | 84 - 85 | 11644 | 85 |
NM_005529.6 | NP_005520.4 | 84 - 85 | 11641 | 85 | |
XM_011541318.2 | XP_011539620.1 | 89 - 90 | 12125 | 85 | |
XM_017001120.1 | XP_016856609.1 | 85 - 86 | 11771 | 85 | |
XM_017001121.1 | XP_016856610.1 | 84 - 85 | 11720 | 85 | |
XM_017001122.1 | XP_016856611.1 | 84 - 85 | 11717 | 85 | |
GenBank mRNA | AB209851.1 | - | 36 - 37 | 5381 | 85 |
BC109204.1 | - | 3 - 4 | 267 | 85 | |
M85289.1 | - | 84 - 85 | 11641 | 85 | |
X62515.1 | - | 84 - 85 | 11607 | 85 |