The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015570.3 | NP_056385.1 | 12 - 13 | 2640 | 72 |
XM_005250257.2 | XP_005250314.1 | 5 - 6 | 714 | 72 | |
XM_011516010.2 | XP_011514312.1 | 12 - 13 | 3105 | 72 | |
XM_011516011.2 | XP_011514313.1 | 12 - 13 | 3102 | 72 | |
XM_011516012.2 | XP_011514314.1 | 11 - 12 | 3039 | 72 | |
XM_011516017.2 | XP_011514319.1 | 11 - 12 | 11659 | 72 | |
XM_011516018.2 | XP_011514320.1 | 12 - 13 | 1640 | 72 | |
XM_017011951.1 | XP_016867440.1 | 12 - 13 | 3105 | 72 | |
GenBank mRNA | AF326917.1 | - | 12 - 13 | 2226 | 72 |
AK296981.1 | - | 4 - 5 | 310 | 72 |