Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LASP1
Entrez Gene ID
3927
Gene Name
LIM and SH3 protein 1
Gene Aliases
Lasp-1, MLN50
Chromosome Location
Chr.17: 38869859 - 38921770 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006148.3 NP_006139.1 3 - 4 578 67
GenBank mRNA AK295522.1 - 3 - 4 376 67
AK296092.1 - 3 - 4 376 67
BC007560.1 - 3 - 4 370 67
CN371189.1 - 3 - 4 331 67
X82456.1 - 3 - 4 322 67

View Full Product Details