The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145294.4 | NP_660337.3 | 10 - 11 | 1174 | 103 |
XM_017023023.1 | XP_016878512.1 | 10 - 11 | 1177 | 103 | |
GenBank mRNA | AB067511.1 | - | 9 - 10 | 1565 | 103 |
AK091062.1 | - | 10 - 11 | 1174 | 103 | |
BC121185.1 | - | 10 - 11 | 1187 | 103 | |
BC121186.1 | - | 10 - 11 | 1184 | 103 |