Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHTF18
Entrez Gene ID
63922
Gene Name
chromosome transmission fidelity factor 18
Gene Aliases
C16orf41, C321D2.2, C321D2.3, C321D2.4, CHL12, Ctf18, RUVBL
Chromosome Location
Chr.16: 788622 - 798074 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022092.2 NP_071375.1 14 - 15 1872 88
XM_005255470.1 XP_005255527.1 14 - 15 1851 88
XM_005255471.3 XP_005255528.1 13 - 14 1954 88
XM_011522572.1 XP_011520874.1 14 - 15 1871 88
XM_011522573.1 XP_011520875.1 6 - 7 831 88
XM_017023532.1 XP_016879021.1 14 - 15 1835 88
XM_017023533.1 XP_016879022.1 14 - 15 1871 88
XM_017023534.1 XP_016879023.1 14 - 15 1863 88
GenBank mRNA AK024476.1 - 11 - 12 3278 88
AK128869.1 - 10 - 11 2881 88
BC006278.2 - 14 - 15 1839 88
BC006437.1 - 13 - 14 1932 88
BC018184.1 - 14 - 15 1847 88

View Full Product Details