Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WDR45
Entrez Gene ID
11152
Gene Name
WD repeat domain 45
Gene Aliases
JM5, NBIA4, NBIA5, WDRX1, WIPI-4, WIPI4
Chromosome Location
Chr.X: 49074433 - 49101121 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029896.1 NP_001025067.1 4 - 5 403 61
GenBank mRNA AJ005897.1 - 4 - 5 359 61
AK225122.1 - 4 - 5 382 61
AY691428.1 - 3 - 4 231 61
BC000464.2 - 4 - 5 397 61
BC003037.1 - 4 - 5 397 61
BQ068083.1 - 4 - 5 403 61
CR456725.1 - 3 - 4 231 61

View Full Product Details