The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001006658.2 | NP_001006659.1 | 4 - 5 | 854 | 79 |
NM_001877.4 | NP_001868.2 | 4 - 5 | 854 | 79 | |
XM_011509206.2 | XP_011507508.1 | 4 - 5 | 750 | 79 | |
GenBank mRNA | AK223627.1 | - | 4 - 5 | 826 | 79 |
AK301496.1 | - | 4 - 5 | 822 | 79 | |
BC090937.1 | - | 4 - 5 | 789 | 79 | |
BC136394.1 | - | 4 - 5 | 828 | 79 | |
J03565.1 | - | 4 - 5 | 798 | 79 | |
M26004.1 | - | 4 - 5 | 804 | 79 | |
Y00649.1 | - | 4 - 5 | 790 | 79 |