The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001006658.2 | NP_001006659.1 | 9 - 10 | 1681 | 86 |
NM_001877.4 | NP_001868.2 | 9 - 10 | 1681 | 86 | |
XM_011509206.2 | XP_011507508.1 | 9 - 10 | 1577 | 86 | |
GenBank mRNA | AK223627.1 | - | 9 - 10 | 1653 | 86 |
AK301496.1 | - | 9 - 10 | 1649 | 86 | |
BC090937.1 | - | 9 - 10 | 1616 | 86 | |
BC136394.1 | - | 9 - 10 | 1655 | 86 | |
J03565.1 | - | 9 - 10 | 1625 | 86 | |
M26004.1 | - | 9 - 10 | 1631 | 86 | |
Y00649.1 | - | 9 - 10 | 1614 | 86 |