Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- TSGA10
- Entrez Gene ID
-
80705
- Gene Name
- testis specific 10
- Gene Aliases
-
CEP4L, CT79
- Chromosome Location
-
Chr.2: 98997261 - 99154962 on Build GRCh38
View Full Product Details