The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142293.1 | NP_001135765.1 | 11 - 12 | 1287 | 89 |
NM_014570.4 | NP_055385.3 | 12 - 13 | 1419 | 89 | |
GenBank mRNA | AF111847.1 | - | 12 - 13 | 1256 | 89 |
AK022768.1 | - | 7 - 8 | 684 | 89 | |
AK297635.1 | - | 10 - 11 | 1069 | 89 | |
AK300531.1 | - | 11 - 12 | 1060 | 89 | |
AK307967.1 | - | 11 - 12 | 1129 | 89 | |
AK315720.1 | - | 12 - 13 | 1257 | 89 | |
AL137598.1 | - | 11 - 12 | 1071 | 89 | |
AL159143.1 | - | 12 - 13 | 1265 | 89 | |
BC005122.2 | - | 12 - 13 | 1228 | 89 | |
CR456382.1 | - | 12 - 13 | 1210 | 89 |