The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005498.3 | NP_001005498.2 | 1 - 2 | 199 | 63 |
NM_024599.5 | NP_078875.4 | 2 - 3 | 274 | 63 | |
XM_005257669.3 | XP_005257726.1 | 2 - 3 | 635 | 63 | |
XM_005257670.1 | XP_005257727.1 | 2 - 3 | 231 | 63 | |
XM_011525249.2 | XP_011523551.1 | 2 - 3 | 1678 | 63 | |
XM_011525250.2 | XP_011523552.1 | 3 - 4 | 388 | 63 | |
XM_011525251.2 | XP_011523553.1 | 2 - 3 | 205 | 63 | |
XM_017025079.1 | XP_016880568.1 | 2 - 3 | 635 | 63 | |
GenBank mRNA | AK292135.1 | - | 2 - 3 | 267 | 63 |
AK292166.1 | - | 1 - 2 | 154 | 63 | |
AY500247.1 | - | 1 - 2 | 50 | 63 | |
BC016034.1 | - | 2 - 3 | 254 | 63 | |
BP284970.1 | - | 2 - 3 | 273 | 63 | |
BP288209.1 | - | 2 - 3 | 273 | 63 | |
DC337840.1 | - | 2 - 3 | 274 | 63 |