The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001009877.2 | NP_001009877.2 | 12 - 13 | 1379 | 67 |
NM_001317951.1 | NP_001304880.1 | 12 - 13 | 1731 | 67 | |
NM_023924.4 | NP_076413.3 | 12 - 13 | 1552 | 67 | |
NR_134296.1 | 12 - 13 | 1888 | 67 | ||
GenBank mRNA | AK023503.1 | - | 12 - 13 | 1709 | 67 |
AK024392.1 | - | 9 - 10 | 3732 | 67 | |
AK026830.1 | - | 7 - 8 | 683 | 67 | |
AK297573.1 | - | 12 - 13 | 1366 | 67 | |
AK299157.1 | - | 12 - 13 | 1552 | 67 | |
AL833925.1 | - | 4 - 5 | 419 | 67 | |
AY358630.1 | - | 2 - 3 | 1075 | 67 | |
BC007217.1 | - | 13 - 14 | 1668 | 67 | |
BC041590.1 | - | 10 - 11 | 1238 | 67 | |
BX648703.1 | - | 12 - 13 | 1890 | 67 | |
DQ248311.1 | - | 8 - 9 | 877 | 67 |