Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TP53I11
Entrez Gene ID
9537
Gene Name
tumor protein p53 inducible protein 11
Gene Aliases
PIG11
Chromosome Location
Chr.11: 44932348 - 44951306 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001258322.1 NP_001245251.1 2 - 3 478 72
NM_001258323.2 NP_001245252.1 2 - 3 305 72
NM_001318384.1 NP_001305313.1 2 - 3 313 72
GenBank mRNA AK056379.1 - 2 - 3 241 72
DA115495.1 - 2 - 3 93 72
DA120301.1 - 2 - 3 136 72

View Full Product Details