The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243137.1 | NP_001230066.1 | 12 - 13 | 1103 | 76 |
NM_002605.2 | NP_002596.1 | 12 - 13 | 1305 | 76 | |
NM_173454.1 | NP_775656.1 | 11 - 12 | 1167 | 76 | |
XM_011521668.2 | XP_011519970.1 | 5 - 6 | 1439 | 76 | |
XM_017022308.1 | XP_016877797.1 | 9 - 10 | 1536 | 76 | |
XM_017022309.1 | XP_016877798.1 | 10 - 11 | 903 | 76 | |
XM_017022310.1 | XP_016877799.1 | 12 - 13 | 1842 | 76 | |
XM_017022311.1 | XP_016877800.1 | 13 - 14 | 2040 | 76 | |
GenBank mRNA | AF056490.1 | - | 10 - 11 | 768 | 76 |
AF332653.1 | - | 12 - 13 | 1252 | 76 | |
AF388183.1 | - | 12 - 13 | 1298 | 76 | |
AF388184.1 | - | 11 - 12 | 1160 | 76 | |
AF388185.1 | - | 10 - 11 | 1081 | 76 | |
AF388186.1 | - | 11 - 12 | 1219 | 76 | |
AF388187.1 | - | 13 - 14 | 1496 | 76 | |
AK074280.1 | - | 12 - 13 | 1103 | 76 | |
AK127232.1 | - | 9 - 10 | 2586 | 76 | |
BC060762.1 | - | 13 - 14 | 1368 | 76 | |
BC075822.1 | - | 12 - 13 | 1196 | 76 |