The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002605.2 | NP_002596.1 | 2 - 3 | 428 | 115 |
NM_173454.1 | NP_775656.1 | 2 - 3 | 428 | 115 | |
XM_017022308.1 | XP_016877797.1 | 2 - 3 | 965 | 115 | |
XM_017022309.1 | XP_016877798.1 | 2 - 3 | 138 | 115 | |
XM_017022310.1 | XP_016877799.1 | 2 - 3 | 965 | 115 | |
XM_017022311.1 | XP_016877800.1 | 2 - 3 | 965 | 115 | |
GenBank mRNA | AF332653.1 | - | 2 - 3 | 375 | 115 |
AF388183.1 | - | 2 - 3 | 421 | 115 | |
AF388184.1 | - | 2 - 3 | 421 | 115 | |
AF388185.1 | - | 2 - 3 | 421 | 115 | |
AF388186.1 | - | 2 - 3 | 421 | 115 | |
AF388187.1 | - | 2 - 3 | 421 | 115 | |
BC060762.1 | - | 3 - 4 | 491 | 115 | |
BC075822.1 | - | 2 - 3 | 319 | 115 |