The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243137.1 | NP_001230066.1 | 7 - 8 | 707 | 124 |
NM_002605.2 | NP_002596.1 | 7 - 8 | 909 | 124 | |
XM_017022309.1 | XP_016877798.1 | 5 - 6 | 507 | 124 | |
XM_017022310.1 | XP_016877799.1 | 7 - 8 | 1446 | 124 | |
GenBank mRNA | AF056490.1 | - | 5 - 6 | 372 | 124 |
AF332653.1 | - | 7 - 8 | 856 | 124 | |
AF388183.1 | - | 7 - 8 | 902 | 124 | |
AK074280.1 | - | 7 - 8 | 707 | 124 | |
BC060762.1 | - | 8 - 9 | 972 | 124 | |
BC075822.1 | - | 7 - 8 | 800 | 124 |