Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
E2F8
Entrez Gene ID
79733
Gene Name
E2F transcription factor 8
Gene Aliases
E2F-8
Chromosome Location
Chr.11: 19224063 - 19241655 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256371.1 NP_001243300.1 5 - 6 1085 74
NM_001256372.1 NP_001243301.1 5 - 6 1030 74
NM_024680.3 NP_078956.2 5 - 6 1284 74
GenBank mRNA AK055206.1 - 5 - 6 1226 74
AK292688.1 - 5 - 6 995 74
BC126400.1 - 5 - 6 1275 74
BC126402.1 - 5 - 6 1275 74
BU164108.1 - 3 - 4 634 74

View Full Product Details