The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014716.3 | NP_055531.1 | 16 - 17 | 1711 | 64 |
GenBank mRNA | BC018543.1 | - | 16 - 17 | 1686 | 64 |
BT009788.1 | - | 16 - 17 | 1505 | 64 | |
D30758.1 | - | 16 - 17 | 1693 | 64 |