The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024708.3 | NP_078984.2 | 3 - 4 | 719 | 90 |
NM_198243.2 | NP_937886.1 | 3 - 4 | 719 | 90 | |
GenBank mRNA | AF451994.1 | - | 3 - 4 | 639 | 90 |
AK026204.1 | - | 3 - 4 | 639 | 90 | |
AK222958.1 | - | 3 - 4 | 639 | 90 | |
AK222974.1 | - | 3 - 4 | 639 | 90 | |
AK289860.1 | - | 3 - 4 | 672 | 90 | |
BC063581.1 | - | 3 - 4 | 660 | 90 |