Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TRIM66
Entrez Gene ID
9866
Gene Name
tripartite motif containing 66
Gene Aliases
C11orf29, TIF1D, TIF1DELTA
Chromosome Location
Chr.11: 8612037 - 8683217 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014818.1 NP_055633.1 20 - 20 9557 132
XM_006718397.2 XP_006718460.1 25 - 25 10495 132
XM_006718398.3 XP_006718461.1 21 - 21 11107 132
XM_011520504.1 XP_011518806.1 26 - 26 10538 132
XM_011520507.2 XP_011518809.1 25 - 25 10459 132
XM_011520508.1 XP_011518810.1 25 - 25 10538 132
XM_011520509.1 XP_011518811.1 24 - 24 10420 132
XM_011520510.1 XP_011518812.1 25 - 25 10411 132
XM_011520511.1 XP_011518813.1 25 - 25 10384 132
XM_011520512.1 XP_011518814.1 21 - 21 9759 132
XM_011520513.1 XP_011518815.1 21 - 21 9757 132
XM_011520514.2 XP_011518816.1 21 - 21 9713 132
XM_011520515.1 XP_011518817.1 22 - 22 9926 132
XM_011520516.1 XP_011518818.1 22 - 22 9926 132
XM_011520517.2 XP_011518819.1 22 - 22 11248 132
XM_011520518.1 XP_011518820.1 22 - 22 9896 132
XM_011520519.1 XP_011518821.1 21 - 21 9784 132
XM_011520522.2 XP_011518824.1 20 - 20 10962 132
XM_011520523.2 XP_011518825.1 21 - 21 11844 132
XM_011520526.2 XP_011518828.1 13 - 13 8813 132
XM_011520527.1 XP_011518829.1 13 - 13 8081 132
XM_017018629.1 XP_016874118.1 25 - 25 10888 132
XM_017018630.1 XP_016874119.1 20 - 20 9850 132
XM_017018631.1 XP_016874120.1 20 - 20 10002 132
GenBank mRNA AK000908.1 - 1 - 1 1304 132
BC031354.1 - 1 - 1 806 132

View Full Product Details