The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177478.1 | NP_001170949.1 | 4 - 5 | 1404 | 68 |
NM_001177479.1 | NP_001170950.1 | 5 - 6 | 1551 | 68 | |
NM_144657.4 | NP_653258.2 | 4 - 5 | 1442 | 68 | |
XM_006724619.1 | XP_006724682.1 | 3 - 4 | 1295 | 68 | |
XM_011530863.2 | XP_011529165.1 | 5 - 6 | 1624 | 68 | |
XM_011530864.1 | XP_011529166.1 | 5 - 6 | 1531 | 68 | |
XM_017029274.1 | XP_016884763.1 | 4 - 5 | 1422 | 68 | |
GenBank mRNA | AK055240.1 | - | 4 - 5 | 1378 | 68 |
BC101720.1 | - | 4 - 5 | 1354 | 68 | |
BC101722.1 | - | 4 - 5 | 1354 | 68 | |
BC143522.1 | - | 5 - 6 | 1463 | 68 | |
BX538112.1 | - | 4 - 5 | 1414 | 68 | |
BX649166.1 | - | 4 - 5 | 1417 | 68 |