The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001143993.1 | NP_001137465.1 | 3 - 4 | 894 | 127 |
NM_001143994.1 | NP_001137466.1 | 3 - 4 | 859 | 127 | |
NM_003475.3 | NP_003466.1 | 3 - 4 | 859 | 127 | |
XM_005253149.3 | XP_005253206.2 | 4 - 5 | 1010 | 127 | |
XM_005253150.1 | XP_005253207.1 | 3 - 4 | 921 | 127 | |
XM_006718338.2 | XP_006718401.1 | 3 - 4 | 1262 | 127 | |
XM_011520389.1 | XP_011518691.1 | 3 - 4 | 916 | 127 | |
XM_017018360.1 | XP_016873849.1 | 4 - 5 | 1010 | 127 | |
XM_017018361.1 | XP_016873850.1 | 3 - 4 | 921 | 127 | |
XM_017018362.1 | XP_016873851.1 | 3 - 4 | 921 | 127 | |
GenBank mRNA | BC106921.1 | - | 3 - 4 | 867 | 127 |
BC106922.1 | - | 3 - 4 | 867 | 127 | |
M91083.1 | - | 3 - 4 | 867 | 127 |