The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198075.3 | NP_932341.1 | 8 - 9 | 1130 | 77 |
XM_005252775.2 | XP_005252832.1 | 8 - 9 | 1954 | 77 | |
XM_005252776.2 | XP_005252833.1 | 8 - 9 | 1954 | 77 | |
XM_006718132.2 | XP_006718195.1 | 8 - 9 | 2029 | 77 | |
XM_006718133.2 | XP_006718196.1 | 7 - 8 | 837 | 77 | |
XM_011519875.2 | XP_011518177.1 | 9 - 10 | 1369 | 77 | |
XM_011519876.1 | XP_011518178.1 | 9 - 10 | 2025 | 77 | |
XM_011519877.2 | XP_011518179.1 | 8 - 9 | 1106 | 77 | |
XM_017017167.1 | XP_016872656.1 | 9 - 10 | 1444 | 77 | |
XM_017017168.1 | XP_016872657.1 | 9 - 10 | 1683 | 77 | |
GenBank mRNA | AK024495.1 | - | 7 - 8 | 2597 | 77 |
BC016439.1 | - | 6 - 7 | 664 | 77 | |
BC035937.1 | - | 8 - 9 | 1116 | 77 |