Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RBFA
Entrez Gene ID
79863
Gene Name
ribosome binding factor A (putative)
Gene Aliases
C18orf22, HsT169
Chromosome Location
Chr.18: 80034346 - 80050652 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024805.2 NP_079081.2 5 - 6 733 110
GenBank mRNA AF261073.2 - 5 - 6 613 110
AK024825.1 - 5 - 6 721 110
BC014195.2 - 5 - 6 595 110
BC057772.1 - 5 - 6 601 110

View Full Product Details