The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319072.1 | NP_001306001.1 | 10 - 11 | 1022 | 106 |
NM_001319073.1 | NP_001306002.1 | 12 - 13 | 1190 | 106 | |
NM_014016.4 | NP_054735.3 | 11 - 12 | 1150 | 106 | |
XM_011533500.2 | XP_011531802.1 | 11 - 12 | 941 | 106 | |
GenBank mRNA | AB020658.1 | - | 11 - 12 | 994 | 106 |
AK291142.1 | - | 11 - 12 | 1034 | 106 | |
AK296421.1 | - | 10 - 11 | 804 | 106 | |
AK301248.1 | - | 12 - 13 | 1030 | 106 | |
AL136831.1 | - | 11 - 12 | 940 | 106 | |
BC016559.1 | - | 11 - 12 | 946 | 106 | |
CR749564.1 | - | 11 - 12 | 2096 | 106 |