The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319072.1 | NP_001306001.1 | 11 - 12 | 1099 | 142 |
NM_001319073.1 | NP_001306002.1 | 13 - 14 | 1267 | 142 | |
NM_014016.4 | NP_054735.3 | 12 - 13 | 1227 | 142 | |
XM_011533500.2 | XP_011531802.1 | 12 - 13 | 1018 | 142 | |
GenBank mRNA | AB020658.1 | - | 12 - 13 | 1071 | 142 |
AK291142.1 | - | 12 - 13 | 1111 | 142 | |
AK296421.1 | - | 11 - 12 | 881 | 142 | |
AK301248.1 | - | 13 - 14 | 1107 | 142 | |
AL136831.1 | - | 12 - 13 | 1017 | 142 | |
BC016559.1 | - | 12 - 13 | 1023 | 142 | |
CR749564.1 | - | 12 - 13 | 2173 | 142 |