Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GPSM2
Entrez Gene ID
29899
Gene Name
G-protein signaling modulator 2
Gene Aliases
CMCS, DFNB82, LGN, PINS
Chromosome Location
Chr.1: 108875350 - 108934335 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001321038.1 NP_001307967.1 9 - 10 1319 97
NM_001321039.1 NP_001307968.1 9 - 10 1551 97
NM_013296.4 NP_037428.3 9 - 10 1551 97
XM_006710589.2 XP_006710652.1 8 - 9 2010 97
XM_011541301.2 XP_011539603.1 10 - 11 1828 97
XM_011541302.2 XP_011539604.1 10 - 11 2490 97
XM_011541303.2 XP_011539605.1 9 - 10 2246 97
XM_017001097.1 XP_016856586.1 10 - 11 2396 97
XM_017001098.1 XP_016856587.1 10 - 11 2164 97
GenBank mRNA AB445462.1 - 9 - 10 1551 97
AK295563.1 - 9 - 10 1138 97
AK314249.1 - 9 - 10 1547 97
AY136740.1 - 8 - 9 1034 97
BC027732.2 - 9 - 10 1327 97
CR456786.1 - 8 - 9 1034 97
U54999.1 - 8 - 9 1207 97

View Full Product Details