Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GPSM2
Entrez Gene ID
29899
Gene Name
G-protein signaling modulator 2
Gene Aliases
CMCS, DFNB82, LGN, PINS
Chromosome Location
Chr.1: 108875350 - 108934335 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011541302.2 XP_011539604.1 2 - 3 1193 88

View Full Product Details