The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199739.1 | NP_001186668.1 | 5 - 6 | 906 | 87 |
NM_004079.4 | NP_004070.3 | 6 - 7 | 1056 | 87 | |
GenBank mRNA | AK226095.1 | - | 6 - 7 | 918 | 87 |
AK301472.1 | - | 5 - 6 | 741 | 87 | |
AK314482.1 | - | 6 - 7 | 897 | 87 | |
BC002642.2 | - | 6 - 7 | 857 | 87 | |
CR541676.1 | - | 5 - 6 | 795 | 87 | |
M86553.1 | - | 5 - 6 | 801 | 87 | |
M90696.1 | - | 6 - 7 | 931 | 87 | |
S93414.1 | - | 5 - 6 | 801 | 87 |