The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004475.2 | NP_004466.2 | 7 - 8 | 824 | 62 |
XM_005257950.3 | XP_005258007.1 | 7 - 8 | 855 | 62 | |
XM_005257953.2 | XP_005258010.1 | 6 - 7 | 759 | 62 | |
XM_017024394.1 | XP_016879883.1 | 7 - 8 | 752 | 62 | |
XM_017024395.1 | XP_016879884.1 | 8 - 9 | 921 | 62 | |
XM_017024396.1 | XP_016879885.1 | 7 - 8 | 758 | 62 | |
GenBank mRNA | BC003683.1 | - | 7 - 8 | 736 | 62 |
BC017292.1 | - | 6 - 7 | 646 | 62 | |
BT019477.1 | - | 5 - 6 | 554 | 62 | |
BT019478.1 | - | 5 - 6 | 554 | 62 | |
CR541784.1 | - | 5 - 6 | 554 | 62 | |
CR542265.1 | - | 5 - 6 | 554 | 62 | |
M60922.1 | - | 7 - 8 | 680 | 62 |