Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VPS45
Entrez Gene ID
11311
Gene Name
vacuolar protein sorting 45 homolog
Gene Aliases
H1, H1VPS45, SCN5, VPS45A, VPS45B, VPS54A, VSP45, VSP45A
Chromosome Location
Chr.1: 150067293 - 150145327 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001279353.1 NP_001266282.1 11 - 12 1501 138
NM_001279354.1 NP_001266283.1 13 - 14 1697 138
NM_001279355.1 NP_001266284.1 10 - 11 1308 138
NM_007259.4 NP_009190.2 13 - 14 2052 138
GenBank mRNA AF165513.1 - 13 - 14 1532 138
AJ133421.1 - 13 - 14 1520 138
AK023170.1 - 13 - 14 1539 138
AK223214.1 - 13 - 14 1505 138
AK295529.1 - 13 - 14 1552 138
AK298786.1 - 11 - 12 1412 138
AK298835.1 - 10 - 11 1168 138
AY453586.1 - 7 - 8 1391 138
BC012932.1 - 13 - 14 1538 138
BC028382.2 - 13 - 14 2033 138

View Full Product Details