Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VPS45
Entrez Gene ID
11311
Gene Name
vacuolar protein sorting 45 homolog
Gene Aliases
H1, H1VPS45, SCN5, VPS45A, VPS45B, VPS54A, VSP45, VSP45A
Chromosome Location
Chr.1: 150067293 - 150145327 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007259.4 NP_009190.2 1 - 2 662 106
NR_103998.1 1 - 2 287 106
XM_017000164.1 XP_016855653.1 1 - 2 212 106
GenBank mRNA AF165513.1 - 1 - 2 142 106
AJ133421.1 - 1 - 2 130 106
AK023170.1 - 1 - 2 149 106
AK223214.1 - 1 - 2 115 106
AK293555.1 - 1 - 2 148 106
AK307949.1 - 1 - 2 108 106
BC012932.1 - 1 - 2 148 106
BC028382.2 - 1 - 2 643 106
DA374440.1 - 1 - 2 287 106
U35246.1 - 1 - 2 140 106

View Full Product Details