The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001279354.1 | NP_001266283.1 | 5 - 6 | 643 | 79 |
NM_007259.4 | NP_009190.2 | 5 - 6 | 998 | 79 | |
XM_017000164.1 | XP_016855653.1 | 5 - 6 | 548 | 79 | |
GenBank mRNA | AF165513.1 | - | 5 - 6 | 478 | 79 |
AJ133421.1 | - | 5 - 6 | 466 | 79 | |
AK023170.1 | - | 5 - 6 | 485 | 79 | |
AK223214.1 | - | 5 - 6 | 451 | 79 | |
AK295529.1 | - | 5 - 6 | 498 | 79 | |
AK298835.1 | - | 2 - 3 | 114 | 79 | |
AK301994.1 | - | 5 - 6 | 605 | 79 | |
BC012932.1 | - | 5 - 6 | 484 | 79 | |
BC028382.2 | - | 5 - 6 | 979 | 79 |