The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004698.2 | NP_004689.1 | 12 - 13 | 1801 | 111 |
XM_011510129.1 | XP_011508431.1 | 13 - 14 | 1843 | 111 | |
XM_011510130.2 | XP_011508432.1 | 9 - 10 | 2363 | 111 | |
XM_017002790.1 | XP_016858279.1 | 9 - 10 | 1237 | 111 | |
GenBank mRNA | AF001947.1 | - | 11 - 12 | 1697 | 111 |
AF016370.1 | - | 12 - 13 | 1711 | 111 | |
AK222825.1 | - | 12 - 13 | 1727 | 111 | |
AK297139.1 | - | 12 - 13 | 1685 | 111 | |
AK297265.1 | - | 11 - 12 | 1580 | 111 | |
AK299980.1 | - | 13 - 14 | 1818 | 111 | |
AK310241.1 | - | 10 - 11 | 1468 | 111 | |
AK312891.1 | - | 12 - 13 | 1722 | 111 | |
BC000184.2 | - | 11 - 12 | 1688 | 111 | |
BC001954.1 | - | 11 - 12 | 1696 | 111 | |
BQ002848.1 | - | 4 - 5 | 111 |