The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004698.2 | NP_004689.1 | 4 - 5 | 583 | 58 |
XM_011510129.1 | XP_011508431.1 | 5 - 6 | 625 | 58 | |
XM_011510131.2 | XP_011508433.1 | 4 - 5 | 523 | 58 | |
XM_011510132.2 | XP_011508434.1 | 4 - 5 | 523 | 58 | |
XM_017002791.1 | XP_016858280.1 | 4 - 5 | 523 | 58 | |
GenBank mRNA | AF016370.1 | - | 4 - 5 | 493 | 58 |
AK222825.1 | - | 4 - 5 | 509 | 58 | |
AK297139.1 | - | 4 - 5 | 467 | 58 | |
AK299980.1 | - | 5 - 6 | 605 | 58 | |
AK312891.1 | - | 4 - 5 | 504 | 58 | |
BC000184.2 | - | 3 - 4 | 470 | 58 | |
BC001954.1 | - | 3 - 4 | 478 | 58 |