Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VPS39
Entrez Gene ID
23339
Gene Name
VPS39, HOPS complex subunit
Gene Aliases
TLP, VAM6, hVam6p
Chromosome Location
Chr.15: 42158701 - 42208331 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001301138.1 NP_001288067.1 1 - 2 241 98
NM_015289.3 NP_056104.2 1 - 2 241 98
XM_011521403.2 XP_011519705.1 1 - 2 210 98
XM_011521404.2 XP_011519706.1 1 - 2 246 98
XM_017022036.1 XP_016877525.1 1 - 2 209 98
XM_017022037.1 XP_016877526.1 1 - 2 246 98
XM_017022038.1 XP_016877527.1 1 - 2 224 98
XM_017022039.1 XP_016877528.1 1 - 2 214 98
GenBank mRNA AB018313.2 - 1 - 2 192 98
AF280814.1 - 1 - 2 68 98
AF281052.1 - 1 - 2 211 98
BC068559.1 - 1 - 2 210 98
BX537404.1 - 1 - 2 220 98
DB461120.1 - 1 - 2 242 98

View Full Product Details