The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301138.1 | NP_001288067.1 | 23 - 24 | 2536 | 79 |
NM_015289.3 | NP_056104.2 | 22 - 23 | 2503 | 79 | |
XM_011521403.2 | XP_011519705.1 | 23 - 24 | 2505 | 79 | |
XM_011521404.2 | XP_011519706.1 | 22 - 23 | 2508 | 79 | |
XM_017022036.1 | XP_016877525.1 | 23 - 24 | 2504 | 79 | |
XM_017022037.1 | XP_016877526.1 | 22 - 23 | 2508 | 79 | |
XM_017022038.1 | XP_016877527.1 | 22 - 23 | 2691 | 79 | |
XM_017022039.1 | XP_016877528.1 | 22 - 23 | 2502 | 79 | |
GenBank mRNA | AB018313.2 | - | 22 - 23 | 2454 | 79 |
AF280814.1 | - | 23 - 24 | 2363 | 79 | |
AF281052.1 | - | 22 - 23 | 2473 | 79 | |
AK025334.1 | - | 7 - 8 | 703 | 79 | |
AK128403.1 | - | 5 - 6 | 1825 | 79 | |
BC015817.2 | - | 18 - 19 | 2063 | 79 | |
BC068559.1 | - | 22 - 23 | 2472 | 79 | |
BX537404.1 | - | 22 - 23 | 2482 | 79 |