The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301138.1 | NP_001288067.1 | 8 - 9 | 739 | 87 |
NM_015289.3 | NP_056104.2 | 7 - 8 | 706 | 87 | |
XM_011521403.2 | XP_011519705.1 | 8 - 9 | 708 | 87 | |
XM_011521404.2 | XP_011519706.1 | 7 - 8 | 711 | 87 | |
XM_017022036.1 | XP_016877525.1 | 8 - 9 | 707 | 87 | |
XM_017022037.1 | XP_016877526.1 | 7 - 8 | 711 | 87 | |
XM_017022038.1 | XP_016877527.1 | 7 - 8 | 894 | 87 | |
XM_017022039.1 | XP_016877528.1 | 7 - 8 | 679 | 87 | |
GenBank mRNA | AB018313.2 | - | 7 - 8 | 657 | 87 |
AF280814.1 | - | 8 - 9 | 566 | 87 | |
AF281052.1 | - | 7 - 8 | 676 | 87 | |
BC015817.2 | - | 3 - 4 | 266 | 87 | |
BC068559.1 | - | 7 - 8 | 675 | 87 |