Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TEX10
Entrez Gene ID
54881
Gene Name
testis expressed 10
Gene Aliases
Ipi1, bA208F1.2
Chromosome Location
Chr.9: 100302075 - 100352977 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017746.3 NP_060216.2 1 - 2 206 142
XM_011518798.1 XP_011517100.1 1 - 2 168 142
XM_017014848.1 XP_016870337.1 1 - 2 168 142
GenBank mRNA AB060968.1 - 1 - 2 122 142
AK000294.1 - 1 - 2 122 142
AK298916.1 - 1 - 2 125 142
BP336313.1 - 1 - 2 206 142

View Full Product Details